missing translation for 'onlineSavingsMsg'
Learn More

2-Bromonicotinic Acid 98.0+%, TCI America™

Catalog No. B26265G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B26265G 5 g
B26261G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B26265G Supplier TCI America Supplier No. B26265G

Chemical Identifiers

CAS 35905-85-2
Molecular Formula C6H4BrNO2
Molecular Weight (g/mol) 202.007
MDL Number MFCD01646068
InChI Key MMWNKXIFVYQOTK-UHFFFAOYSA-N
Synonym 2-bromonicotinic acid, 2-bromo-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 2-bromo, 2-bromo-nicotinic acid, 2-bromo-3-carboxypyridine, 3-bromopyridine-3-carboxylic acid, bromo-nicotinic acid, pubchem5071, acmc-1aj4p, ksc223s8f
PubChem CID 233262
IUPAC Name 2-bromopyridine-3-carboxylic acid
SMILES C1=CC(=C(N=C1)Br)C(=O)O

Specifications

Melting Point 254°C
Quantity 5 g
Formula Weight 202.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromonicotinic Acid