missing translation for 'onlineSavingsMsg'
Learn More

2-Bromonicotinic acid, 97%

CAS: 35905-85-2 | C6H4BrNO2 | 202.007 g/mol

$236.55 - $236.55

Chemical Identifiers

CAS 35905-85-2
Molecular Formula C6H4BrNO2
Molecular Weight (g/mol) 202.007
MDL Number MFCD01646068
InChI Key MMWNKXIFVYQOTK-UHFFFAOYSA-N
Synonym 2-bromonicotinic acid, 2-bromo-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 2-bromo, 2-bromo-nicotinic acid, 2-bromo-3-carboxypyridine, 3-bromopyridine-3-carboxylic acid, bromo-nicotinic acid, pubchem5071, acmc-1aj4p, ksc223s8f
PubChem CID 233262
IUPAC Name 2-bromopyridine-3-carboxylic acid
SMILES C1=CC(=C(N=C1)Br)C(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH3023906
View Documents
Thermo Scientific Chemicals
H3023906
5 g
Each for $236.55
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

35905-85-2
202.007
MMWNKXIFVYQOTK-UHFFFAOYSA-N
233262
C1=CC(=C(N=C1)Br)C(=O)O
C6H4BrNO2
MFCD01646068
2-bromonicotinic acid, 2-bromo-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 2-bromo, 2-bromo-nicotinic acid, 2-bromo-3-carboxypyridine, 3-bromopyridine-3-carboxylic acid, bromo-nicotinic acid, pubchem5071, acmc-1aj4p, ksc223s8f
2-bromopyridine-3-carboxylic acid

Specifications

35905-85-2
C6H4BrNO2
5 g
MMWNKXIFVYQOTK-UHFFFAOYSA-N
2-bromopyridine-3-carboxylic acid
233262
97%
∼254°C
MFCD01646068
2-bromonicotinic acid, 2-bromo-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 2-bromo, 2-bromo-nicotinic acid, 2-bromo-3-carboxypyridine, 3-bromopyridine-3-carboxylic acid, bromo-nicotinic acid, pubchem5071, acmc-1aj4p, ksc223s8f
C1=CC(=C(N=C1)Br)C(=O)O
202.007
202.01
2-Bromonicotinic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only