missing translation for 'onlineSavingsMsg'
Learn More

2-Bromonicotinaldehyde 98.0+%, TCI America™

Catalog No. B26251G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B26251G 1 g
B26255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B26251G Supplier TCI America Supplier No. B26251G

Chemical Identifiers

CAS 128071-75-0
Molecular Formula C6H4BrNO
Molecular Weight (g/mol) 186.008
InChI Key GNFWMEFWZWXLIN-UHFFFAOYSA-N
Synonym 2-bromonicotinaldehyde, 2-bromo-3-formylpyridine, 2-bromo-3-pyridinecarboxaldehyde, 2-bromopyridine-3-carboxaldehyde, 3-pyridinecarboxaldehyde, 2-bromo, pubchem5057, acmc-1bypv, 2-bromo-3-formyl pyridine, bromo-3-pyridine carbaldehyde, bromo-3-pyridinecarboxaldehyde
PubChem CID 4714951
IUPAC Name 2-bromopyridine-3-carbaldehyde
SMILES C1=CC(=C(N=C1)Br)C=O

Specifications

Melting Point 73°C
Boiling Point 100°C
Quantity 1 g
Formula Weight 186.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromonicotinaldehyde