missing translation for 'onlineSavingsMsg'
Learn More

2-Bromonaphthalene 98.0+%, TCI America™

Catalog No. B06195G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B06195G 5 g
B061925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B06195G Supplier TCI America Supplier No. B06195G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 580-13-2
Molecular Formula C10H7Br
Molecular Weight (g/mol) 207.07
MDL Number MFCD00004051
InChI Key APSMUYYLXZULMS-UHFFFAOYSA-N
Synonym naphthalene, 2-bromo, 2-naphthyl bromide, 2-bromo-naphthalene, beta-bromonaphthalene, beta-naphthyl bromide, .beta.-bromonaphthalene, unii-syu33i753n, naphthalene,2-bromo, .beta.-naphthyl bromide, pbn-2
PubChem CID 11372
IUPAC Name 2-bromonaphthalene
SMILES BrC1=CC=C2C=CC=CC2=C1

Specifications

Melting Point 58°C
Color White-Yellow
Boiling Point 282°C
Quantity 5 g
Formula Weight 207.07
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromonaphthalene