missing translation for 'onlineSavingsMsg'
Learn More

2-Bromonaphthalene, 98+%

Catalog No. AAA1530006
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1530006 5 g
AAA1530014 25 g
AAA1530022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1530006 Supplier Thermo Scientific Chemicals Supplier No. A1530006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 580-13-2 | C10H7Br | 207.07 g/mol

2-Bromonaphthalene is used as a raw material in the preparation of biaryls through Suzuki cross coupling reaction. Further, it plays a vital role in the preparation of dyes. It is also used to study potential tumorigenicity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromonaphthalene is used as a raw material in the preparation of biaryls through Suzuki cross coupling reaction. Further, it plays a vital role in the preparation of dyes. It is also used to study potential tumorigenicity.

Solubility
Slightly soluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 580-13-2
Molecular Formula C10H7Br
Molecular Weight (g/mol) 207.07
MDL Number MFCD00004051
InChI Key APSMUYYLXZULMS-UHFFFAOYSA-N
PubChem CID 11372
IUPAC Name 2-bromonaphthalene
SMILES BrC1=CC=C2C=CC=CC2=C1

Specifications

Melting Point 53°C to 57°C
Density 1.605
Boiling Point 281°C to 282°C
Flash Point 112°C (233°F)
Odor Odorless
Quantity 5 g
Beilstein 1858110
Merck Index 14,1426
Synonym naphthalene, 2-bromo, 2-naphthyl bromide, 2-bromo-naphthalene, beta-bromonaphthalene, beta-naphthyl bromide, .beta.-bromonaphthalene, unii-syu33i753n, naphthalene,2-bromo, .beta.-naphthyl bromide, pbn-2
Solubility Information Slightly soluble in water.
Formula Weight 207.08
Percent Purity ≥98%
Chemical Name or Material 2-Bromonaphthalene
Show More Show Less
Hazard Category H302-H319
Hazard Statement GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280-P301+P312-P305+P351+P338-P330-P501c
EINECSNumber 209-452-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only