missing translation for 'onlineSavingsMsg'
Learn More

2-(Bromomethyl)pyridine Hydrobromide 98.0+%, TCI America™

Catalog No. B37725G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37725G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B37725G Supplier TCI America Supplier No. B37725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 31106-82-8
Molecular Formula C6H7Br2N
Molecular Weight (g/mol) 252.937
MDL Number MFCD01863544
InChI Key JQDNCGRNPYKRAO-UHFFFAOYSA-N
Synonym 2-bromomethyl pyridine hydrobromide, 2-picolyl bromide hydrobromide, 2-bromomethyl pyridine hbr, pyridine, 2-bromomethyl-, hydrobromide, 2-bromomethyl pyridine, bromide, c6h6brn.hbr, acmc-20aim5, ksc495g6r, 2-bromomethylpyridine hydrobromide, bromomethyl pyridine hydrobromide
PubChem CID 2734720
IUPAC Name 2-(bromomethyl)pyridine;hydrobromide
SMILES C1=CC=NC(=C1)CBr.Br

Specifications

Melting Point 149°C
Color Yellow
Quantity 5 g
UN Number 3261
Formula Weight 252.94
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Bromomethyl)pyridine Hydrobromide