missing translation for 'onlineSavingsMsg'
Learn More

2-(Bromomethyl)oxetane, 96%

CAS: 939759-23-6 | C4H7BrO | 151.00 g/mol

$609.03 - $1,865.93

Chemical Identifiers

CAS 939759-23-6
Molecular Formula C4H7BrO
Molecular Weight (g/mol) 151.00
MDL Number MFCD08544405
InChI Key OMXAGUVERXNCSZ-UHFFFAOYNA-N
Synonym 2-bromomethyl oxetane, 2-bromomethyl-oxetane, oxetane, 2-bromomethyl, cyclobutane, bromomethyl, 2-bromomethyl oxetane oxetane, 2-bromomethyl
PubChem CID 16244494
IUPAC Name 2-(bromomethyl)oxetane
SMILES BrCC1CCO1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH66276MD
View Documents
Thermo Scientific Chemicals
H66276MD
250 mg N/A
Only null left
 
AAH6627603
View Documents
Thermo Scientific Chemicals
H6627603
1 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

939759-23-6
151.00
OMXAGUVERXNCSZ-UHFFFAOYNA-N
16244494
BrCC1CCO1
C4H7BrO
MFCD08544405
2-bromomethyl oxetane, 2-bromomethyl-oxetane, oxetane, 2-bromomethyl, cyclobutane, bromomethyl, 2-bromomethyl oxetane oxetane, 2-bromomethyl
2-(bromomethyl)oxetane

Specifications

939759-23-6
C4H7BrO
250 mg
2-bromomethyl oxetane, 2-bromomethyl-oxetane, oxetane, 2-bromomethyl, cyclobutane, bromomethyl, 2-bromomethyl oxetane oxetane, 2-bromomethyl
BrCC1CCO1
151.00
151
Liquid
Colorless to Yellow
MFCD08544405
Moisture sensitive
OMXAGUVERXNCSZ-UHFFFAOYNA-N
2-(bromomethyl)oxetane
16244494
96%
2-(Bromomethyl)oxetane
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only