Learn More
2-(Bromomethyl)-6-pyrrolidin-1-ylpyridine, 97%, Thermo Scientific™
Supplier:  Thermo Scientific Chemicals CC38008CB
 View more versions of this product
Catalog No. CC38008CB
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Specifications
Specifications
2-(Bromomethyl)-6-pyrrolidin-1-ylpyridine
96.5
97%
C10H13BrN2
250mg
UFXKRINLXMWRES-UHFFFAOYSA-N
2-(bromomethyl)-6-pyrrolidin-1-ylpyridine
24229482
97%
869901-04-2
100.0
Amber Glass Bottle
MFCD09817465
2-bromomethyl-6-pyrrolidin-1-yl pyridine, 2-bromomethyl-6-pyrrolidin-1-ylpyridine, pyridine,2-bromomethyl-6-1-pyrrolidinyl, 6-bromomethyl-2-pyrrolidinylpyridine, 1-6-bromomethyl pyridin-2-yl pyrrolidine, pyridine, 2-bromomethyl-6-1-pyrrolidinyl
C1CCN(C1)C2=CC=CC(=N2)CBr
241.132
241.13
Chemical Identifiers
869901-04-2
241.132
UFXKRINLXMWRES-UHFFFAOYSA-N
24229482
C1CCN(C1)C2=CC=CC(=N2)CBr
C10H13BrN2
MFCD09817465
2-bromomethyl-6-pyrrolidin-1-yl pyridine, 2-bromomethyl-6-pyrrolidin-1-ylpyridine, pyridine,2-bromomethyl-6-1-pyrrolidinyl, 6-bromomethyl-2-pyrrolidinylpyridine, 1-6-bromomethyl pyridin-2-yl pyrrolidine, pyridine, 2-bromomethyl-6-1-pyrrolidinyl
2-(bromomethyl)-6-pyrrolidin-1-ylpyridine
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
SDS Documents