missing translation for 'onlineSavingsMsg'
Learn More

2-(Bromomethyl)-2-(hydroxymethyl)-1,3-propanediol 98.0+%, TCI America™

Catalog No. p-7123321
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40571G 1 g
B40575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B40571G Supplier TCI America Supplier No. B40571G
Only null left

Chemical Identifiers

CAS 19184-65-7
Molecular Formula C5H11BrO3
Molecular Weight (g/mol) 199.044
MDL Number MFCD00135306
InChI Key MMHHBAUIJVTLFZ-UHFFFAOYSA-N
Synonym 2-bromomethyl-2-hydroxymethyl propane-1,3-diol, 2-bromomethyl-2-hydroxymethyl-1,3-propanediol, 1,3-propanediol, 2-bromomethyl-2-hydroxymethyl, tris hydroxymethyl bromomethyl methane, monobromoneopentyltriol, 2-bromoethylidynetrimethanol, pentaerythritol monobromohydrin, 1, 2-bromomethyl-2-hydroxymethyl, 2,2-bis hydroxymethyl-3-bromopropan-1-ol, 2,2-bis hydroxymethyl-3-hydroxypropyl bromide
PubChem CID 87954
IUPAC Name 2-(bromomethyl)-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)CBr)O

Specifications

Melting Point 77°C
Color White
Quantity 1 g
Formula Weight 199.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Bromomethyl)-2-(hydroxymethyl)-1,3-propanediol