missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromoisobutyramide 98.0+%, TCI America™
$46.60 - $145.40
Chemical Identifiers
| CAS | 7462-74-0 |
|---|---|
| Molecular Formula | C4H8BrNO |
| Molecular Weight (g/mol) | 166.018 |
| MDL Number | MFCD08234837 |
| InChI Key | WTRIMJTZOOLIFZ-UHFFFAOYSA-N |
| Synonym | 2-Bromo-2-methylpropionamide |
| PubChem CID | 101235 |
| IUPAC Name | 2-bromo-2-methylpropanamide |
| SMILES | CC(C)(C(=O)N)Br |
Chemical Identifiers
| 7462-74-0 | |
| 166.018 | |
| WTRIMJTZOOLIFZ-UHFFFAOYSA-N | |
| 101235 | |
| CC(C)(C(=O)N)Br |
| C4H8BrNO | |
| MFCD08234837 | |
| 2-Bromo-2-methylpropionamide | |
| 2-bromo-2-methylpropanamide |
Specifications
| 7462-74-0 | |
| White-Yellow | |
| MFCD08234837 | |
| 2-Bromo-2-methylpropionamide | |
| CC(C)(C(=O)N)Br | |
| 166.018 | |
| 166.02 | |
| Crystalline Powder |
| 147°C | |
| C4H8BrNO | |
| 5 g | |
| WTRIMJTZOOLIFZ-UHFFFAOYSA-N | |
| 2-bromo-2-methylpropanamide | |
| 101235 | |
| ≥98.0% (GC,N) | |
| 2-Bromoisobutyramide |
Safety and Handling
TSCA : No