missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoindene 98.0+%, TCI America™

Catalog No. b31635g
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31635G 5 g
B316325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B31635G Supplier TCI America Supplier No. B31635G
Only null left

Chemical Identifiers

CAS 10485-09-3
Molecular Formula C9H7Br
Molecular Weight (g/mol) 195.059
MDL Number MFCD06797863
InChI Key CCUYEVNCRQDQRF-UHFFFAOYSA-N
Synonym 2-bromoindene, 1h-indene, 2-bromo, indene, 2-bromo, 2-bromanyl-1h-indene, 2-bromoindene, 95+%, 2-bromo-indene, pubchem9657, #, acmc-1c5rn, sodium perborate,tetrahydrate
PubChem CID 575586
IUPAC Name 2-bromo-1H-indene
SMILES C1C2=CC=CC=C2C=C1Br

Specifications

Melting Point 38°C
Boiling Point 127°C
Quantity 5 g
Formula Weight 195.06
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromoindene
TSCA No
Recommended Storage Refrigerator