missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoimidazole, 95%

Catalog No. p-7034854
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5255303 1 g
AAH5255306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5255303 Supplier Thermo Scientific Chemicals Supplier No. H5255303
Only null left

CAS: 16681-56-4 | C3H3BrN2 | 146.98 g/mol

2-Bromoimidazole is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromoimidazole is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.

Chemical Identifiers

CAS 16681-56-4
Molecular Formula C3H3BrN2
Molecular Weight (g/mol) 146.98
MDL Number MFCD02179526
InChI Key AXHRGVJWDJDYPO-UHFFFAOYSA-N
Synonym 2-bromoimidazole, 1h-imidazole, 2-bromo, bromoimidazole, sftheabililuh@, pubchem8987, zlchem 1065, acmc-1btox, 2-bromanyl-1h-imidazole, ksc176e6b, 2-bromo-1h-imidazole
PubChem CID 2773261
IUPAC Name 2-bromo-1H-imidazole
SMILES BrC1=NC=CN1

Specifications

Melting Point 197°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 146.98
Percent Purity 95%
Chemical Name or Material 2-Bromoimidazole
Hazard Category H302-H315-H318-H335
Hazard Statement GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only