missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2-Bromoethyl)phosphonic Acid 98.0+%, TCI America™
$260.97 - $874.24
Chemical Identifiers
| CAS | 999-82-6 |
|---|---|
| Molecular Formula | C2H6BrO3P |
| Molecular Weight (g/mol) | 188.95 |
| MDL Number | MFCD00013957 |
| InChI Key | BFKXVERGWXHHIH-UHFFFAOYSA-N |
| PubChem CID | 3613627 |
| IUPAC Name | (2-bromoethyl)phosphonic acid |
| SMILES | OP(O)(=O)CCBr |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B5013200MG
|
TCI America
B5013200MG |
200 mg |
Each for $260.97
|
|
|||||
|
B50131G
|
TCI America
B50131G |
1 g |
Each for $874.24
|
|
|||||
Chemical Identifiers
| 999-82-6 | |
| 188.95 | |
| BFKXVERGWXHHIH-UHFFFAOYSA-N | |
| (2-bromoethyl)phosphonic acid |
| C2H6BrO3P | |
| MFCD00013957 | |
| 3613627 | |
| OP(O)(=O)CCBr |
Specifications
| 999-82-6 | |
| C2H6BrO3P | |
| 200 mg | |
| BFKXVERGWXHHIH-UHFFFAOYSA-N | |
| (2-bromoethyl)phosphonic acid | |
| 3613627 | |
| ≥98.0% (T) | |
| (2-Bromoethyl)phosphonic Acid |
| 88°C | |
| MFCD00013957 | |
| 3261 | |
| OP(O)(=O)CCBr | |
| 188.95 | |
| 188.94 | |
| Crystalline Powder |
Safety and Handling
TSCA : No