missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoethyl methyl ether, 96%, stab. with sodium carbonate

Catalog No. p-7022542
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1537006 5 g
AAA1537014 25 g
AAA1537022 100 g
AAA1537036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1537006 Supplier Thermo Scientific Chemicals Supplier No. A1537006
Only null left

CAS: 6482-24-2 | C3H7BrO | 138.992 g/mol

2-Bromoethyl methyl ether has been used as starting reagent in the synthesis of 1-bromo-4-(2-methoxyethoxy)benzene1 and 1-bromo-4-[(2-methoxyethoxy)methyl]benzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromoethyl methyl ether has been used as starting reagent in the synthesis of 1-bromo-4-(2-methoxyethoxy)benzene1 and 1-bromo-4-[(2-methoxyethoxy)methyl]benzene.

Solubility
Soluble in water.14,4 g/L at 25°C

Notes
Protect from heat. Incompatible with heat, strong bases and oxidizing agents. Store at 2-8°C.

Chemical Identifiers

CAS 6482-24-2
Molecular Formula C3H7BrO
Molecular Weight (g/mol) 138.992
MDL Number MFCD00000236
InChI Key YZUPZGFPHUVJKC-UHFFFAOYSA-N
Synonym 2-bromoethyl methyl ether, 2-methoxyethyl bromide, ethane, 1-bromo-2-methoxy, ether, 2-bromoethyl methyl, methoxyethyl bromide, 2-bromoethylmethylether, bromoethyl methyl ether, 2-bromoethylmethyl ether, 2-bromoethylmethyl-ether, 2-bromo-1-methoxyethane
PubChem CID 80972
IUPAC Name 1-bromo-2-methoxyethane
SMILES COCCBr

Specifications

Density 1.478
Boiling Point 109°C to 110°C
Flash Point 28°C (82°F)
Odor Recognizable
Refractive Index 1.448
Quantity 5 g
UN Number UN3271
Solubility Information Soluble in water.14,4g/L at 25°C
Formula Weight 139
Percent Purity 96%
Chemical Name or Material 2-Bromoethyl methyl ether, stabilized with sodium carbonate
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.
EINECSNumber 229-339-4
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only