missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoethyl methyl ether, 95%

Catalog No. AC402980050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC402980050 5 g Glass bottle
AC402980250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC402980050 Supplier Thermo Scientific Chemicals Supplier No. 402980050

CAS: 6482-24-2 | C3H7BrO | 138.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6482-24-2
Molecular Formula C3H7BrO
Molecular Weight (g/mol) 138.99
MDL Number MFCD00000236
InChI Key YZUPZGFPHUVJKC-UHFFFAOYSA-N
Synonym 2-bromoethyl methyl ether, 2-methoxyethyl bromide, ethane, 1-bromo-2-methoxy, ether, 2-bromoethyl methyl, methoxyethyl bromide, 2-bromoethylmethylether, bromoethyl methyl ether, 2-bromoethylmethyl ether, 2-bromoethylmethyl-ether, 2-bromo-1-methoxyethane
PubChem CID 80972
IUPAC Name 1-bromo-2-methoxyethane
SMILES COCCBr

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.4600g/mL
Boiling Point 107.0°C to 110.0°C
Flash Point 28°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4460 to 1.4500
Linear Formula BrCH2CH2OCH3
Quantity 5 g
Specific Gravity 1.46
Solubility Information Solubility in water: 14.4g/L (25°C)
Formula Weight 138.99
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2-Bromoethyl methyl ether
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN: Wash with plenty of soap a
EINECSNumber 229-339-4
Recommended Storage Product darkens during storage

RUO – Research Use Only