missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoethyl ether, 90%

Catalog No. p-4440177
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC445860050 5 mL
AC445860250 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC445860050 Supplier Thermo Scientific Chemicals Supplier No. 445860050

CAS: 5414-19-7 | C4H8Br2O | 231.92 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5414-19-7
Molecular Formula C4H8Br2O
Molecular Weight (g/mol) 231.92
InChI Key FOZVXADQAHVUSV-UHFFFAOYSA-N
Synonym bis 2-bromoethyl ether, 1-bromo-2-2-bromoethoxy ethane, 2-bromoethyl ether, 2,2'-dibromodiethyl ether, ethane, 1,1'-oxybis 2-bromo, ether, bis 2-bromoethyl, beta,beta'-dibromodiethyl ether, 2,2-dibromodiethyl ether, 2-bromoethylether
PubChem CID 21521
IUPAC Name 1-bromo-2-(2-bromoethoxy)ethane
SMILES C(CBr)OCCBr

Specifications

Density 1.8450g/mL
Boiling Point 92°C to 93°C (12.0 mmHg)
Flash Point 85°C
Assay Percent Range 88% min. (GC)
Packaging Glass Bottle
Linear Formula [Br(CH2)2]2O
Quantity 5 mL
Specific Gravity 1.845
Formula Weight 231.92
Percent Purity 90%
Chemical Name or Material 2-Bromoethyl ether
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 226-504-2

RUO – Research Use Only