missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2-Bromoethyl)diphenylsulfonium Trifluoromethanesulfonate 98.0+%, TCI America™
$96.94 - $313.16
Chemical Identifiers
| CAS | 247129-85-7 |
|---|---|
| Molecular Formula | C15H14BrF3O3S2 |
| Molecular Weight (g/mol) | 443.293 |
| MDL Number | MFCD12546049 |
| InChI Key | SAXNFOVPRAUGNK-UHFFFAOYSA-M |
| Synonym | (2-Bromoethyl)diphenylsulfonium Triflate |
| PubChem CID | 25193077 |
| IUPAC Name | 2-bromoethyl(diphenyl)sulfanium;trifluoromethanesulfonate |
| SMILES | C1=CC=C(C=C1)[S+](CCBr)C2=CC=CC=C2.C(F)(F)(F)S(=O)(=O)[O-] |
Chemical Identifiers
| 247129-85-7 | |
| 443.293 | |
| SAXNFOVPRAUGNK-UHFFFAOYSA-M | |
| 25193077 | |
| C1=CC=C(C=C1)[S+](CCBr)C2=CC=CC=C2.C(F)(F)(F)S(=O)(=O)[O-] |
| C15H14BrF3O3S2 | |
| MFCD12546049 | |
| (2-Bromoethyl)diphenylsulfonium Triflate | |
| 2-bromoethyl(diphenyl)sulfanium;trifluoromethanesulfonate |
Specifications
| 247129-85-7 | |
| White | |
| MFCD12546049 | |
| (2-Bromoethyl)diphenylsulfonium Triflate | |
| C1=CC=C(C=C1)[S+](CCBr)C2=CC=CC=C2.C(F)(F)(F)S(=O)(=O)[O-] | |
| 443.293 | |
| 443.29 | |
| Crystalline Powder |
| 90°C | |
| C15H14BrF3O3S2 | |
| 1 g | |
| SAXNFOVPRAUGNK-UHFFFAOYSA-M | |
| 2-bromoethyl(diphenyl)sulfanium;trifluoromethanesulfonate | |
| 25193077 | |
| ≥98.0% (HPLC,T) | |
| (2-Bromoethyl)diphenylsulfonium Trifluoromethanesulfonate |
Safety and Handling
TSCA : No