missing translation for 'onlineSavingsMsg'
Learn More

(2-Bromoethyl)diphenylsulfonium Trifluoromethanesulfonate 98.0+%, TCI America™

Catalog No. B39581G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B39581G 1 g
B39585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B39581G Supplier TCI America Supplier No. B39581G
Only null left

Chemical Identifiers

CAS 247129-85-7
Molecular Formula C15H14BrF3O3S2
Molecular Weight (g/mol) 443.293
MDL Number MFCD12546049
InChI Key SAXNFOVPRAUGNK-UHFFFAOYSA-M
Synonym (2-Bromoethyl)diphenylsulfonium Triflate
PubChem CID 25193077
IUPAC Name 2-bromoethyl(diphenyl)sulfanium;trifluoromethanesulfonate
SMILES C1=CC=C(C=C1)[S+](CCBr)C2=CC=CC=C2.C(F)(F)(F)S(=O)(=O)[O-]

Specifications

Melting Point 90°C
Color White
Quantity 1 g
Formula Weight 443.29
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (2-Bromoethyl)diphenylsulfonium Trifluoromethanesulfonate