missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoethyl acrylate, 94%, stab. with 900-1500ppm 4-methoxyphenol

Catalog No. p-7045928
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1250203 1 g
AAL1250206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1250203 Supplier Thermo Scientific Chemicals Supplier No. L1250203
Only null left

CAS: 4823-47-6 | C5H7BrO2 | 179.013 g/mol

2-Bromoethyl acrylate is utilized in curable and reactive polymers due to the presence of reactive bromine. It is also used in the synthesis of other monomers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromoethyl acrylate is utilized in curable and reactive polymers due to the presence of reactive bromine. It is also used in the synthesis of other monomers.

Solubility
Miscible with water.

Notes
Light and heat sensitive Incompatible with strong oxidizing agents, polymerizing agents and peroxides.

Chemical Identifiers

CAS 4823-47-6
Molecular Formula C5H7BrO2
Molecular Weight (g/mol) 179.013
MDL Number MFCD00013541
InChI Key CDZAAIHWZYWBSS-UHFFFAOYSA-N
PubChem CID 20963
IUPAC Name 2-bromoethyl prop-2-enoate
SMILES C=CC(=O)OCCBr

Specifications

Boiling Point 52°C to 53°C (5 mmHg)
Quantity 1 g
Beilstein 1749713
Synonym 2-bromoethyl acrylate, acrylic acid, 2-bromoethyl ester, 2-propenoic acid, 2-bromoethyl ester, ccris 4753, acrylic acid 2-bromoethyl ester, 2-bromoethylacrylate, 2-bromoethyl=acrylate, acmc-20ao1p, acrylic acid 2-bromo-ethyl ester, 2-propenoic acid,2-bromoethyl ester
Solubility Information Miscible with water.
Formula Weight 179.01
Percent Purity 94%
Chemical Name or Material 2-Bromoethyl acrylate, stabilized with 0.1% 4-methoxyphenol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber AS4810000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only