missing translation for 'onlineSavingsMsg'
Learn More

(2-Bromoethoxy)-tert-butyldimethylsilane, 98+%, stabilized, Thermo Scientific Chemicals

Catalog No. AC399830100
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC399830100 10 g Glass bottle
AC399830010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC399830100 Supplier Thermo Scientific Chemicals Supplier No. 399830100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 86864-60-0 | Molecular Formula: C8H19BrOSi | Molecular Weight: 239.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 86864-60-0
Molecular Formula C8H19BrOSi
Molecular Weight (g/mol) 239.22
MDL Number MFCD00209550
InChI Key JBKINHFZTVLNEM-UHFFFAOYSA-N
Synonym 2-bromoethoxy-tert-butyldimethylsilane, 2-bromoethoxy tert-butyl dimethylsilane, 2-bromoethoxy-t-butyl dimethylsilane, 2-t-butyldimethylsiloxy ethylbromide, 2-bromoethoxy-t-butyldimethylsilane, 2-bromo-ethoxy-tert-butyl-dimethyl-silane, silane, 2-bromoethoxy 1,1-dimethylethyl dimethyl, brch2ch2otbs
PubChem CID 3608067
IUPAC Name 2-bromoethoxy-tert-butyl-dimethylsilane
SMILES CC(C)(C)[Si](C)(C)OCCBr

Specifications

CAS Min % 98.0
CAS Max % 100.0
Color White to Yellow
Density 1.1150g/mL
Boiling Point 70°C to 75°C (2.5 mmHg)
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)CSi(CH3)2O(CH2)2Br
Quantity 10 g
Specific Gravity 1.115
Stabilizer 5 % max. Na2CO3
Formula Weight 239.22
Percent Purity 98+%
Physical Form Crystalline Solid or Crystals
Chemical Name or Material (2-Bromoethoxy)-tert-butyldimethylsilane
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only