missing translation for 'onlineSavingsMsg'
Learn More

(2-Bromoethoxy)-tert-butyldimethylsilane, 98%, stab. with sodium carbonate, Thermo Scientific Chemicals

Catalog No. AAH5659403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5659403 1 g
AAH5659406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5659403 Supplier Thermo Scientific Chemicals Supplier No. H5659403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 86864-60-0
Molecular Formula C8H19BrOSi
Molecular Weight (g/mol) 239.228
MDL Number MFCD00209550
InChI Key JBKINHFZTVLNEM-UHFFFAOYSA-N
Synonym 2-bromoethoxy-tert-butyldimethylsilane, 2-bromoethoxy tert-butyl dimethylsilane, 2-bromoethoxy-t-butyl dimethylsilane, 2-t-butyldimethylsiloxy ethylbromide, 2-bromoethoxy-t-butyldimethylsilane, 2-bromo-ethoxy-tert-butyl-dimethyl-silane, silane, 2-bromoethoxy 1,1-dimethylethyl dimethyl, brch2ch2otbs
PubChem CID 3608067
IUPAC Name 2-bromoethoxy-tert-butyl-dimethylsilane
SMILES CC(C)(C)[Si](C)(C)OCCBr

Specifications

Boiling Point 120°C (100mmHg)
Color Colorless
Physical Form Liquid
Quantity 1 g
Flash Point 73°C (163°F)
Formula Weight 239.23
Percent Purity 98%
Refractive Index 1.444
Density 1.115 g/mL
Chemical Name or Material (2-Bromoethoxy)-tert-butyldimethylsilane, Stabilized with sodium carbonate
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only