missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoethanol 95.0+%, TCI America™

Catalog No. p-7118547
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B059025G 25 g
B0590100G 100 g
B0590500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B059025G Supplier TCI America Supplier No. B059025G
Only null left

Chemical Identifiers

CAS 540-51-2
Molecular Formula C2H5BrO
Molecular Weight (g/mol) 124.965
MDL Number MFCD00002827
InChI Key LDLCZOVUSADOIV-UHFFFAOYSA-N
Synonym ethylene bromohydrin, ethanol, 2-bromo, 2-bromoethan-1-ol, glycol bromohydrin, 2-hydroxyethyl bromide, 2-bromo-1-ethanol, 1-bromo-2-ethanol, ethylenebromohydrin, 2-bromoethyl alcohol, 2-bromo-ethanol
PubChem CID 10898
IUPAC Name 2-bromoethanol
SMILES C(CBr)O

Specifications

Melting Point -80°C
Color Yellow
Boiling Point 150°C
Quantity 25 g
UN Number 2922
Formula Weight 124.97
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromoethanol
EINECSNumber (2)-2002
RTECSNumber KJ8225000
TSCA Yes
Recommended Storage Freezer