missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoethanol, 97%

Catalog No. AC106920050
Change view
Click to view available options
Quantity:
5 mL
100 mL
250 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC106920050 5 mL Glass bottle
AC106921000 100 mL Glass bottle
AC106922500 250 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC106920050 Supplier Thermo Scientific Chemicals Supplier No. 106920050

CAS: 540-51-2 | C2H5BrO | 124.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 540-51-2
Molecular Formula C2H5BrO
Molecular Weight (g/mol) 124.96
MDL Number MFCD00002827
InChI Key LDLCZOVUSADOIV-UHFFFAOYSA-N
Synonym ethylene bromohydrin, ethanol, 2-bromo, 2-bromoethan-1-ol, glycol bromohydrin, 2-hydroxyethyl bromide, 2-bromo-1-ethanol, 1-bromo-2-ethanol, ethylenebromohydrin, 2-bromoethyl alcohol, 2-bromo-ethanol
PubChem CID 10898
IUPAC Name 2-bromoethanol
SMILES C(CBr)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -80.0°C
Color Colorless to Yellow
Density 1.7600g/mL
Boiling Point 149.0°C to 150.0°C (750.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4900 to 1.4940
Linear Formula BrCH2CH2OH
Quantity 5 mL
Beilstein 01, 338
Fieser 11,77
Specific Gravity 1.76
Merck Index 15, 3846
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with ethanol, ether, soluble in most common organic solvents,, except petroleum ether
Formula Weight 124.96
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Bromoethanol, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water
EINECSNumber 208-748-1
RTECSNumber KJ8225000
TSCA TSCA
Recommended Storage Product may darken during storage

RUO – Research Use Only