missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoethanesulfonic acid, sodium salt, 98%

Catalog No. AC160030250
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC160030250 25 g
AC160031000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC160030250 Supplier Thermo Scientific Chemicals Supplier No. 160030250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4263-52-9 | C2H4BrNaO3S | 211.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4263-52-9
Molecular Formula C2H4BrNaO3S
Molecular Weight (g/mol) 211.01
MDL Number MFCD00007530
InChI Key HNFOAHXBHLWKNF-UHFFFAOYSA-M
Synonym sodium 2-bromoethanesulfonate, 2-bromoethanesulfonic acid sodium salt, sodium 2-bromoethanesulphonate, 2-bromoethanesulfonic acid, sodium salt, sodium bromoethylsulfonate, ethanesulfonic acid, 2-bromo-, sodium salt, ethanesulfonic acid, 2-bromo-, sodium salt 1:1, sodium 2-bromoethane sulfonate, 2-bromoethanesulfonic acid sodium, natrium-2-bromethansulfonat
PubChem CID 23666797
IUPAC Name sodium;2-bromoethanesulfonate
SMILES C(CBr)S(=O)(=O)[O-].[Na+]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 282.0°C to 286.0°C
Color White
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ex Br) On dry substance (Argentometry)
Packaging Glass bottle
Linear Formula BrCH2CH2SO3Na
Quantity 25 g
Beilstein 04, 7
Solubility Information Solubility in water: in water: 500g/L (20°C)
Formula Weight 211.01
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromoethanesulfonic acid, sodium salt, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 224-244-4
TSCA TSCA

RUO – Research Use Only