missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromodibenzothiophene 95.0+%, TCI America™
$173.37 - $508.89
Chemical Identifiers
| CAS | 22439-61-8 |
|---|---|
| Molecular Formula | C12H7BrS |
| Molecular Weight (g/mol) | 263.152 |
| MDL Number | MFCD00089285 |
| InChI Key | IJICRIUYZZESMW-UHFFFAOYSA-N |
| PubChem CID | 299508 |
| IUPAC Name | 2-bromodibenzothiophene |
| SMILES | C1=CC=C2C(=C1)C3=C(S2)C=CC(=C3)Br |
Chemical Identifiers
| 22439-61-8 | |
| 263.152 | |
| IJICRIUYZZESMW-UHFFFAOYSA-N | |
| 2-bromodibenzothiophene |
| C12H7BrS | |
| MFCD00089285 | |
| 299508 | |
| C1=CC=C2C(=C1)C3=C(S2)C=CC(=C3)Br |
Specifications
| 22439-61-8 | |
| White | |
| MFCD00089285 | |
| 2-bromodibenzo b,d thiophene, dibenzothiophene, 2-bromo, 2-bromobenzo b benzo b thiophene, 4-bromo-8-thiatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene, acmc-209fwi, 2-bromo-dibenzothiophene, dibenzothiophene,2-bromo, 2-bromodibenzo b thiophene, 2-bromo-dibenzo b,d thiophene, dibenzo b,d thiophene, 2-bromo | |
| C1=CC=C2C(=C1)C3=C(S2)C=CC(=C3)Br | |
| 263.152 | |
| 263.15 | |
| Crystalline Powder |
| 125°C | |
| C12H7BrS | |
| 5 g | |
| IJICRIUYZZESMW-UHFFFAOYSA-N | |
| 2-bromodibenzothiophene | |
| 299508 | |
| ≥95.0% (GC) | |
| 2-Bromodibenzothiophene |
Safety and Handling
TSCA : No