missing translation for 'onlineSavingsMsg'
Learn More

2-Bromodibenzothiophene, 98%

CAS: 22439-61-8 | C12H7BrS | 263.152 g/mol

Supplier:  Thermo Scientific Chemicals H6464506

 View more versions of this product

Catalog No. AAH6464506


Only null left
Explore available promotions

Description

Description

Used as intermediate for organic light-emitting diode(OLED) materials and pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as intermediate for organic light-emitting diode(OLED) materials and pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Specifications

2-Bromodibenzothiophene
123°C to 129°C (literature)
MFCD00089285
142203
Slightly soluble in water.
C1=CC=C2C(=C1)C3=C(S2)C=CC(=C3)Br
263.152
263.15
22439-61-8
C12H7BrS
5 g
2-bromodibenzo b,d thiophene, dibenzothiophene, 2-bromo, 2-bromobenzo b benzo b thiophene, 4-bromo-8-thiatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene, acmc-209fwi, 2-bromo-dibenzothiophene, dibenzothiophene,2-bromo, 2-bromodibenzo b thiophene, 2-bromo-dibenzo b,d thiophene, dibenzo b,d thiophene, 2-bromo
IJICRIUYZZESMW-UHFFFAOYSA-N
2-bromodibenzothiophene
299508
98%

Chemical Identifiers

22439-61-8
263.152
IJICRIUYZZESMW-UHFFFAOYSA-N
299508
C1=CC=C2C(=C1)C3=C(S2)C=CC(=C3)Br
C12H7BrS
MFCD00089285
2-bromodibenzo b,d thiophene, dibenzothiophene, 2-bromo, 2-bromobenzo b benzo b thiophene, 4-bromo-8-thiatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene, acmc-209fwi, 2-bromo-dibenzothiophene, dibenzothiophene,2-bromo, 2-bromodibenzo b thiophene, 2-bromo-dibenzo b,d thiophene, dibenzo b,d thiophene, 2-bromo
2-bromodibenzothiophene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only