missing translation for 'onlineSavingsMsg'
Learn More

2-Bromodibenzothiophene, 98%

Catalog No. AAH6464503
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6464503 1 g
AAH6464506 5 g
AAH6464514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6464503 Supplier Thermo Scientific Chemicals Supplier No. H6464503

CAS: 22439-61-8 | C12H7BrS | 263.152 g/mol

Used as intermediate for organic light-emitting diode(OLED) materials and pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as intermediate for organic light-emitting diode(OLED) materials and pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 22439-61-8
Molecular Formula C12H7BrS
Molecular Weight (g/mol) 263.152
MDL Number MFCD00089285
InChI Key IJICRIUYZZESMW-UHFFFAOYSA-N
Synonym 2-bromodibenzo b,d thiophene, dibenzothiophene, 2-bromo, 2-bromobenzo b benzo b thiophene, 4-bromo-8-thiatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene, acmc-209fwi, 2-bromo-dibenzothiophene, dibenzothiophene,2-bromo, 2-bromodibenzo b thiophene, 2-bromo-dibenzo b,d thiophene, dibenzo b,d thiophene, 2-bromo
PubChem CID 299508
IUPAC Name 2-bromodibenzothiophene
SMILES C1=CC=C2C(=C1)C3=C(S2)C=CC(=C3)Br

Specifications

Melting Point 123°C to 129°C (literature)
Quantity 1 g
Beilstein 142203
Solubility Information Slightly soluble in water.
Formula Weight 263.15
Percent Purity 98%
Chemical Name or Material 2-Bromodibenzothiophene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only