missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromocarbazole 98.0+%, TCI America™
$92.41 - $269.52
Chemical Identifiers
| CAS | 3652-90-2 |
|---|---|
| Molecular Formula | C12H8BrN |
| Molecular Weight (g/mol) | 246.11 |
| MDL Number | MFCD09750430 |
| InChI Key | PJRGCJBBXGNEGD-UHFFFAOYSA-N |
| PubChem CID | 12717089 |
| IUPAC Name | 2-bromo-9H-carbazole |
| SMILES | BrC1=CC2=C(C=C1)C1=C(N2)C=CC=C1 |
Chemical Identifiers
| 3652-90-2 | |
| 246.11 | |
| PJRGCJBBXGNEGD-UHFFFAOYSA-N | |
| 2-bromo-9H-carbazole |
| C12H8BrN | |
| MFCD09750430 | |
| 12717089 | |
| BrC1=CC2=C(C=C1)C1=C(N2)C=CC=C1 |
Specifications
| 3652-90-2 | |
| White-Yellow | |
| MFCD09750430 | |
| PJRGCJBBXGNEGD-UHFFFAOYSA-N | |
| 2-bromo-9H-carbazole | |
| 12717089 | |
| ≥98.0% (GC) | |
| 2-Bromocarbazole |
| 255°C | |
| C12H8BrN | |
| 5 g | |
| BrC1=CC2=C(C=C1)C1=C(N2)C=CC=C1 | |
| 246.11 | |
| 246.11 | |
| Crystalline Powder |
Safety and Handling
TSCA : No