missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromobutyric Acid 98.0+%, TCI America™
$42.87 - $236.73
Chemical Identifiers
| CAS | 80-58-0 |
|---|---|
| Molecular Formula | C4H7BrO2 |
| Molecular Weight (g/mol) | 167.00 |
| MDL Number | MFCD00004216,MFCD00210111,MFCD00210112 |
| InChI Key | YAQLSKVCTLCIIE-UHFFFAOYNA-N |
| Synonym | 2-bromobutyric acid, alpha-bromobutyric acid, butanoic acid, 2-bromo, butyric acid, 2-bromo, .alpha.-bromobytyric acid, butyric acid, .alpha.-bromo, dl-2-bromobutyric acid, a-bromobutyric acid, butyric acid, alpha-bromo, bromobutyric acid |
| PubChem CID | 6655 |
| IUPAC Name | 2-bromobutanoic acid |
| SMILES | CCC(Br)C(O)=O |
Chemical Identifiers
| 80-58-0 | |
| 167.00 | |
| YAQLSKVCTLCIIE-UHFFFAOYNA-N | |
| 6655 | |
| CCC(Br)C(O)=O |
| C4H7BrO2 | |
| MFCD00004216,MFCD00210111,MFCD00210112 | |
| 2-bromobutyric acid, alpha-bromobutyric acid, butanoic acid, 2-bromo, butyric acid, 2-bromo, .alpha.-bromobytyric acid, butyric acid, .alpha.-bromo, dl-2-bromobutyric acid, a-bromobutyric acid, butyric acid, alpha-bromo, bromobutyric acid | |
| 2-bromobutanoic acid |
Specifications
| 80-58-0 | |
| Yellow | |
| C4H7BrO2 | |
| 25 g | |
| 2-bromobutyric acid, alpha-bromobutyric acid, butanoic acid, 2-bromo, butyric acid, 2-bromo, .alpha.-bromobytyric acid, butyric acid, .alpha.-bromo, dl-2-bromobutyric acid, a-bromobutyric acid, butyric acid, alpha-bromo, bromobutyric acid | |
| CCC(Br)C(O)=O | |
| 167.00 | |
| 167.00 | |
| Liquid |
| -4°C | |
| 217°C | |
| MFCD00004216,MFCD00210111,MFCD00210112 | |
| 3265 | |
| YAQLSKVCTLCIIE-UHFFFAOYNA-N | |
| 2-bromobutanoic acid | |
| 6655 | |
| ≥98.0% (GC,T) | |
| 2-Bromobutyric Acid |
Safety and Handling
EINECSNumber : (2)-1159
TSCA : Yes