missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobutyric Acid 98.0+%, TCI America™

Catalog No. B0564500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0564500G 500 g
B056425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0564500G Supplier TCI America Supplier No. B0564500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 80-58-0
Molecular Formula C4H7BrO2
Molecular Weight (g/mol) 167.00
MDL Number MFCD00004216,MFCD00210111,MFCD00210112
InChI Key YAQLSKVCTLCIIE-UHFFFAOYNA-N
Synonym 2-bromobutyric acid, alpha-bromobutyric acid, butanoic acid, 2-bromo, butyric acid, 2-bromo, .alpha.-bromobytyric acid, butyric acid, .alpha.-bromo, dl-2-bromobutyric acid, a-bromobutyric acid, butyric acid, alpha-bromo, bromobutyric acid
PubChem CID 6655
IUPAC Name 2-bromobutanoic acid
SMILES CCC(Br)C(O)=O

Specifications

Melting Point -4°C
Color Yellow
Boiling Point 217°C
Quantity 500 g
UN Number 3265
Formula Weight 167.00
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Bromobutyric Acid
EINECSNumber (2)-1159
TSCA Yes