missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobutyric acid, 99%

Catalog No. AC164510050
Change view
Click to view available options
Quantity:
5 mL
250 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC164510050 5 mL Glass bottle
AC164512500 250 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC164510050 Supplier Thermo Scientific Chemicals Supplier No. 164510050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 80-58-0 | C4H7BrO2 | 167.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 80-58-0
Molecular Formula C4H7BrO2
Molecular Weight (g/mol) 167.00
MDL Number MFCD00004216,MFCD00210111,MFCD00210112
InChI Key YAQLSKVCTLCIIE-UHFFFAOYNA-N
Synonym 2-bromobutyric acid, alpha-bromobutyric acid, butanoic acid, 2-bromo, butyric acid, 2-bromo, .alpha.-bromobytyric acid, butyric acid, .alpha.-bromo, dl-2-bromobutyric acid, a-bromobutyric acid, butyric acid, alpha-bromo, bromobutyric acid
PubChem CID 6655
IUPAC Name 2-bromobutanoic acid
SMILES CCC(Br)C(O)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -4°C
Color Yellow
Density 1.5600g/mL
Boiling Point 214°C to 217°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4712 to 1.4732
Linear Formula C2H5CH(Br)CO2H
Quantity 5 mL
Beilstein 02, 281
Specific Gravity 1.56
Merck Index 15, 1420
Solubility Information Solubility in water: 66g/L (20°C). Other solubilities: soluble in alcohol and ether
Formula Weight 167
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Bromobutyric acid, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 201-294-5
TSCA TSCA

RUO – Research Use Only