Learn More
2-Bromobutyric acid, 98%
CAS: 80-58-0 | C4H7BrO2 | 167.00 g/mol
$264.79 - $752.01
Chemical Identifiers
| CAS | 80-58-0 |
|---|---|
| Molecular Formula | C4H7BrO2 |
| Molecular Weight (g/mol) | 167.00 |
| MDL Number | MFCD00004216,MFCD00210111,MFCD00210112 |
| InChI Key | YAQLSKVCTLCIIE-UHFFFAOYNA-N |
| Synonym | 2-bromobutyric acid, alpha-bromobutyric acid, butanoic acid, 2-bromo, butyric acid, 2-bromo, .alpha.-bromobytyric acid, butyric acid, .alpha.-bromo, dl-2-bromobutyric acid, a-bromobutyric acid, butyric acid, alpha-bromo, bromobutyric acid |
| PubChem CID | 6655 |
| IUPAC Name | 2-bromobutanoic acid |
| SMILES | CCC(Br)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2174330
|
Thermo Scientific Chemicals
B2174330 |
250 g |
Each for $264.79
|
|
|||||
|
AAB217430B
|
Thermo Scientific Chemicals
B217430B |
1000 g |
Each for $752.01
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 80-58-0 | |
| 167.00 | |
| YAQLSKVCTLCIIE-UHFFFAOYNA-N | |
| 6655 | |
| CCC(Br)C(O)=O |
| C4H7BrO2 | |
| MFCD00004216,MFCD00210111,MFCD00210112 | |
| 2-bromobutyric acid, alpha-bromobutyric acid, butanoic acid, 2-bromo, butyric acid, 2-bromo, .alpha.-bromobytyric acid, butyric acid, .alpha.-bromo, dl-2-bromobutyric acid, a-bromobutyric acid, butyric acid, alpha-bromo, bromobutyric acid | |
| 2-bromobutanoic acid |
Specifications
| 80-58-0 | |
| 1.567 | |
| >110°C (230°F) | |
| 1.472 | |
| 250 g | |
| 1720950 | |
| 2-bromobutyric acid, alpha-bromobutyric acid, butanoic acid, 2-bromo, butyric acid, 2-bromo, .alpha.-bromobytyric acid, butyric acid, .alpha.-bromo, dl-2-bromobutyric acid, a-bromobutyric acid, butyric acid, alpha-bromo, bromobutyric acid | |
| CCC(Br)C(O)=O | |
| 167.00 | |
| 167.01 | |
| 2-Bromobutyric acid |
| -4°C | |
| 99°C to 103°C (10 mmHg) | |
| C4H7BrO2 | |
| MFCD00004216,MFCD00210111,MFCD00210112 | |
| UN3265 | |
| 14,1413 | |
| YAQLSKVCTLCIIE-UHFFFAOYNA-N | |
| 2-bromobutanoic acid | |
| 6655 | |
| 98% |
Safety and Handling
GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 201-294-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only