missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzothiazole, 99%

Catalog No. AAH6328603
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6328603 1 g
AAH6328606 5 g
AAH6328614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6328603 Supplier Thermo Scientific Chemicals Supplier No. H6328603

CAS: 2516-40-7 | C7H4BrNS | 214.08 g/mol

2-Bromobenzothiazole, is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromobenzothiazole, is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, acids and bases.

Chemical Identifiers

CAS 2516-40-7
Molecular Formula C7H4BrNS
Molecular Weight (g/mol) 214.08
MDL Number MFCD02681887
InChI Key DRLMMVPCYXFPEP-UHFFFAOYSA-N
Synonym 2-bromobenzothiazole, 2-bromobenzo d thiazole, benzothiazole, 2-bromo, dsstox_cid_1498, dsstox_rid_76188, dsstox_gsid_21498, 2-bromobenzthiazole, 2-bromo benzothiazole, 2-bromo-benzothiazole, acmc-20a0ou
PubChem CID 612040
IUPAC Name 2-bromo-1,3-benzothiazole
SMILES C1=CC=C2C(=C1)N=C(S2)Br

Specifications

Melting Point 39°C to 43°C
Color Yellow
Flash Point >110°C (230°F)
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 214.08
Percent Purity 99%
Physical Form Crystalline Solid
Chemical Name or Material 2-Bromobenzothiazole
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only