missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromobenzimidazole 98.0+%, TCI America™
$103.97 - $309.95
Chemical Identifiers
| CAS | 54624-57-6 |
|---|---|
| Molecular Formula | C7H5BrN2 |
| Molecular Weight (g/mol) | 197.035 |
| MDL Number | MFCD00160009 |
| InChI Key | PHPYXVIHDRDPDI-UHFFFAOYSA-N |
| PubChem CID | 2776281 |
| IUPAC Name | 2-bromo-1H-benzimidazole |
| SMILES | C1=CC=C2C(=C1)NC(=N2)Br |
Chemical Identifiers
| 54624-57-6 | |
| 197.035 | |
| PHPYXVIHDRDPDI-UHFFFAOYSA-N | |
| 2-bromo-1H-benzimidazole |
| C7H5BrN2 | |
| MFCD00160009 | |
| 2776281 | |
| C1=CC=C2C(=C1)NC(=N2)Br |
Specifications
| 54624-57-6 | |
| White | |
| MFCD00160009 | |
| 2-bromobenzimidazole, 2-bromo-1h-benzo d imidazole, 2-bromo-1h-1,3-benzodiazole, 2-bromo-1h-benzoimidazole, 1h-benzimidazole, 2-bromo, bromobenzimidazole, bromo-benzimidazole, pubchem16113, acmc-1an7o, 1h-benzimidazole,2-bromo | |
| C1=CC=C2C(=C1)NC(=N2)Br | |
| 197.035 | |
| 197.04 | |
| Crystalline Powder |
| 192°C | |
| C7H5BrN2 | |
| 1 g | |
| PHPYXVIHDRDPDI-UHFFFAOYSA-N | |
| 2-bromo-1H-benzimidazole | |
| 2776281 | |
| ≥98.0% (GC,T) | |
| 2-Bromobenzimidazole |
Safety and Handling
TSCA : No