missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzimidazole 98.0+%, TCI America™

Catalog No. B48381G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48381G 1 g
B48385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B48381G Supplier TCI America Supplier No. B48381G

Chemical Identifiers

CAS 54624-57-6
Molecular Formula C7H5BrN2
Molecular Weight (g/mol) 197.035
MDL Number MFCD00160009
InChI Key PHPYXVIHDRDPDI-UHFFFAOYSA-N
PubChem CID 2776281
IUPAC Name 2-bromo-1H-benzimidazole
SMILES C1=CC=C2C(=C1)NC(=N2)Br

Specifications

Melting Point 192°C
Color White
Quantity 1 g
Synonym 2-bromobenzimidazole, 2-bromo-1h-benzo d imidazole, 2-bromo-1h-1,3-benzodiazole, 2-bromo-1h-benzoimidazole, 1h-benzimidazole, 2-bromo, bromobenzimidazole, bromo-benzimidazole, pubchem16113, acmc-1an7o, 1h-benzimidazole,2-bromo
Formula Weight 197.04
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromobenzimidazole