missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzimidazole, 99%

Catalog No. AAH2618303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2618303 1 g
AAH2618306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2618303 Supplier Thermo Scientific Chemicals Supplier No. H2618303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 54624-57-6 | C7H5BrN2 | 197.035 g/mol

Reactant in the synthesis of substituted benzimidazoles. 2-Bromobenzimidazole is the starting compound in the synthesis of polyhalogenobenzimidazoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant in the synthesis of substituted benzimidazoles. 2-Bromobenzimidazole is the starting compound in the synthesis of polyhalogenobenzimidazoles.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 54624-57-6
Molecular Formula C7H5BrN2
Molecular Weight (g/mol) 197.035
MDL Number MFCD00160009
InChI Key PHPYXVIHDRDPDI-UHFFFAOYSA-N
PubChem CID 2776281
IUPAC Name 2-bromo-1H-benzimidazole
SMILES C1=CC=C2C(=C1)NC(=N2)Br

Specifications

Melting Point 195°C to 201°C
Quantity 1 g
Synonym 2-bromobenzimidazole, 2-bromo-1h-benzo d imidazole, 2-bromo-1h-1,3-benzodiazole, 2-bromo-1h-benzoimidazole, 1h-benzimidazole, 2-bromo, bromobenzimidazole, bromo-benzimidazole, pubchem16113, acmc-1an7o, 1h-benzimidazole,2-bromo
Solubility Information Insoluble in water.
Formula Weight 197.03
Percent Purity 99%
Chemical Name or Material 2-Bromobenzimidazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No

RUO – Research Use Only