missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzenesulfonyl Chloride 97.0+%, TCI America™

Catalog No. p-7119725
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B27025G 5 g
B270225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B27025G Supplier TCI America Supplier No. B27025G
Only null left

Chemical Identifiers

CAS 2905-25-1
Molecular Formula C6H4BrClO2S
Molecular Weight (g/mol) 255.51
MDL Number MFCD00051971
InChI Key VFPWGZNNRSQPBT-UHFFFAOYSA-N
Synonym 2-bromobenzene-1-sulfonyl chloride, 2-bromobenzenesulphonyl chloride, 2-bromosulfonyl chloride, o-bromobenzenesulfonyl chloride, 2-bromophenylsulfonyl chloride, 2-bromobenzenesulfonylchloride, 2-bromobenzenesulphonylchloride, 2-bromophenyl chlorosulfone, 2-bromo-benzenesulfonyl chloride, 2-bromobenzene sulfonyl chloride
PubChem CID 520403
IUPAC Name 2-bromobenzene-1-sulfonyl chloride
SMILES ClS(=O)(=O)C1=CC=CC=C1Br

Specifications

Melting Point 51°C
Boiling Point 117°C
Quantity 5 g
UN Number 3261
Formula Weight 255.51
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromobenzenesulfonyl Chloride