missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzenesulfonyl chloride, 98%

Catalog No. AAA1364506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1364506 5 g
AAA1364514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1364506 Supplier Thermo Scientific Chemicals Supplier No. A1364506

CAS: 2905-25-1 | C6H4BrClO2S | 255.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2905-25-1
Molecular Formula C6H4BrClO2S
Molecular Weight (g/mol) 255.51
MDL Number MFCD00051971
InChI Key VFPWGZNNRSQPBT-UHFFFAOYSA-N
Synonym 2-bromobenzene-1-sulfonyl chloride, 2-bromobenzenesulphonyl chloride, 2-bromosulfonyl chloride, o-bromobenzenesulfonyl chloride, 2-bromophenylsulfonyl chloride, 2-bromobenzenesulfonylchloride, 2-bromobenzenesulphonylchloride, 2-bromophenyl chlorosulfone, 2-bromo-benzenesulfonyl chloride, 2-bromobenzene sulfonyl chloride
PubChem CID 520403
SMILES ClS(=O)(=O)C1=CC=CC=C1Br

Specifications

Melting Point 48°C to 52°C
Boiling Point 127°C to 128°C (2.4 mmHg)
Quantity 5 g
UN Number UN3261
Beilstein 2692202
Sensitivity Moisture sensitive
IUPAC Name 2-bromobenzenesulfonyl chloride
Formula Weight 255.52
Percent Purity 98%
Chemical Name or Material 2-Bromobenzenesulfonyl chloride
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only