missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzenesulfonyl chloride, 97%

CAS: 2905-25-1 | C6H4BrClO2S | 255.51 g/mol

$232.32 - $232.32

Chemical Identifiers

CAS 2905-25-1
Molecular Formula C6H4BrClO2S
Molecular Weight (g/mol) 255.51
MDL Number MFCD00051971
InChI Key VFPWGZNNRSQPBT-UHFFFAOYSA-N
Synonym 2-bromobenzene-1-sulfonyl chloride, 2-bromobenzenesulphonyl chloride, 2-bromosulfonyl chloride, o-bromobenzenesulfonyl chloride, 2-bromophenylsulfonyl chloride, 2-bromobenzenesulfonylchloride, 2-bromobenzenesulphonylchloride, 2-bromophenyl chlorosulfone, 2-bromo-benzenesulfonyl chloride, 2-bromobenzene sulfonyl chloride
PubChem CID 520403
IUPAC Name 2-bromobenzenesulfonyl chloride
SMILES ClS(=O)(=O)C1=CC=CC=C1Br
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC306440050
SDS View Documents
Thermo Scientific Chemicals
306440050
5 g Glass bottle
Each for $232.32
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2905-25-1
255.51
VFPWGZNNRSQPBT-UHFFFAOYSA-N
520403
ClS(=O)(=O)C1=CC=CC=C1Br
C6H4BrClO2S
MFCD00051971
2-bromobenzene-1-sulfonyl chloride, 2-bromobenzenesulphonyl chloride, 2-bromosulfonyl chloride, o-bromobenzenesulfonyl chloride, 2-bromophenylsulfonyl chloride, 2-bromobenzenesulfonylchloride, 2-bromobenzenesulphonylchloride, 2-bromophenyl chlorosulfone, 2-bromo-benzenesulfonyl chloride, 2-bromobenzene sulfonyl chloride
2-bromobenzenesulfonyl chloride

Specifications

2905-25-1
100.0
Yellow
96% min. (ex Cl) (Argentometry)
C6H4BrClO2S
MFCD00051971
2-bromobenzene-1-sulfonyl chloride, 2-bromobenzenesulphonyl chloride, 2-bromosulfonyl chloride, o-bromobenzenesulfonyl chloride, 2-bromophenylsulfonyl chloride, 2-bromobenzenesulfonylchloride, 2-bromobenzenesulphonylchloride, 2-bromophenyl chlorosulfone, 2-bromo-benzenesulfonyl chloride, 2-bromobenzene sulfonyl chloride
VFPWGZNNRSQPBT-UHFFFAOYSA-N
2-bromobenzenesulfonyl chloride
520403
97%
2-Bromobenzenesulfonyl chloride, 97%
96.0
50.0°C to 51.0°C
Authentic
Glass bottle
BrC6H4SO2Cl
5 g
Solubility in water: decomposes
ClS(=O)(=O)C1=CC=CC=C1Br
255.51
255.52
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only