missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoanisole, 97%, Thermo Scientific™

Catalog No. p-71551
Change view
Click to view available options
Quantity:
25mL
100mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC170260250 25mL Glass bottle
AC170261000 100mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC170260250 Supplier Thermo Scientific Chemicals Supplier No. 170260250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 578-57-4
Molecular Formula C7H7BrO
Molecular Weight (g/mol) 187.04
MDL Number MFCD00000064
InChI Key HTDQSWDEWGSAMN-UHFFFAOYSA-N
Synonym 2-bromoanisole, o-bromoanisole, benzene, 1-bromo-2-methoxy, 2-methoxybromobenzene, o-anisyl bromide, anisole, o-bromo, o-methoxybromobenzene, 2-bromoanisol, o-methoxyphenyl bromide, 2-methoxyphenyl bromide
PubChem CID 11358
IUPAC Name 1-bromo-2-methoxybenzene
SMILES COC1=CC=CC=C1Br

Specifications

CAS Min % 96.0
CAS Max % 100.0
Density 1.5000g/mL
Boiling Point 223°C
Flash Point 96°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5730 to 1.575
Linear Formula BrC6H4OCH3
Quantity 25mL
Beilstein 06, 197
Specific Gravity 1.5
Solubility Information Solubility in water: immiscible
Formula Weight 187.04
Percent Purity 97%
Chemical Name or Material 2-Bromoanisole, 97%
Show More Show Less
EINECSNumber 209-425-8
RTECSNumber BZ8500000
TSCA TSCA

RUO – Research Use Only