missing translation for 'onlineSavingsMsg'
Learn More

(2-Bromoallyl)trimethylsilane 90.0+%, TCI America™

Catalog No. B16951G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16951G 1 g
B16955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B16951G Supplier TCI America Supplier No. B16951G
Only null left

Chemical Identifiers

CAS 81790-10-5
Molecular Formula C6H13BrSi
Molecular Weight (g/mol) 193.159
MDL Number MFCD00043054
InChI Key LUPQCAARZVEFMT-UHFFFAOYSA-N
PubChem CID 4173130
IUPAC Name 2-bromoprop-2-enyl(trimethyl)silane
SMILES C[Si](C)(C)CC(=C)Br

Specifications

Color Yellow
Quantity 1 g
UN Number 1993
Formula Weight 193.16
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material (2-Bromoallyl)trimethylsilane
TSCA No
Recommended Storage Freezer