missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromoallyl Alcohol 97.0+%, TCI America™
$188.27 - $592.76
Chemical Identifiers
| CAS | 598-19-6 |
|---|---|
| Molecular Formula | C3H5BrO |
| Molecular Weight (g/mol) | 136.976 |
| MDL Number | MFCD09265077 |
| InChI Key | MDFFZNIQPLKQSG-UHFFFAOYSA-N |
| Synonym | 2-Bromo-2-propen-1-ol |
| PubChem CID | 446627 |
| ChEBI | CHEBI:49514 |
| IUPAC Name | 2-bromoprop-2-en-1-ol |
| SMILES | C=C(CO)Br |
Chemical Identifiers
| 598-19-6 | |
| 136.976 | |
| MDFFZNIQPLKQSG-UHFFFAOYSA-N | |
| 446627 | |
| 2-bromoprop-2-en-1-ol |
| C3H5BrO | |
| MFCD09265077 | |
| 2-Bromo-2-propen-1-ol | |
| CHEBI:49514 | |
| C=C(CO)Br |
Specifications
| 598-19-6 | |
| C3H5BrO | |
| 1 g | |
| MDFFZNIQPLKQSG-UHFFFAOYSA-N | |
| 2-bromoprop-2-en-1-ol | |
| 446627 | |
| 136.98 | |
| Liquid |
| 150°C | |
| MFCD09265077 | |
| 2-Bromo-2-propen-1-ol | |
| C=C(CO)Br | |
| 136.976 | |
| CHEBI:49514 | |
| ≥97.0% (GC) | |
| 2-Bromoallyl Alcohol |
Safety and Handling
TSCA : No