missing translation for 'onlineSavingsMsg'
Learn More

2-(Bromoacetyl)naphthalene, 98%

Catalog No. AAB2396009
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2396009 10 g
AAB2396018 50 g
AAB2396030 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2396009 Supplier Thermo Scientific Chemicals Supplier No. B2396009
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 613-54-7 | C12H9BrO | 249.107 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 613-54-7
Molecular Formula C12H9BrO
Molecular Weight (g/mol) 249.107
MDL Number MFCD00004109
InChI Key YHXHHGDUANVQHE-UHFFFAOYSA-N
Synonym 2-bromo-2'-acetonaphthone, bromomethyl 2-naphthyl ketone, 2-bromoacetyl naphthalene, 2-bromo-1-naphthalen-2-yl ethanone, 2-bromo-1-2-naphthyl-1-ethanone, ethanone, 2-bromo-1-2-naphthalenyl, 2-bromo-1-2-naphthyl ethanone, 2-bromo-1-naphthalen-2-yl ethan-1-one, alpha-bromo-2'-acetonaphthone, 2-bromo-1-naphthalen-2-yl-ethanone
PubChem CID 69179
IUPAC Name 2-bromo-1-naphthalen-2-ylethanone
SMILES C1=CC=C2C=C(C=CC2=C1)C(=O)CBr

Specifications

Melting Point 80°C to 85°C
Quantity 10 g
UN Number UN3261
Beilstein 639153
Formula Weight 249.11
Percent Purity 98%
Chemical Name or Material 2-(Bromoacetyl)naphthalene
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 210-348-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only