missing translation for 'onlineSavingsMsg'
Learn More

2'-Bromoacetophenone 98.0+%, TCI America™

Catalog No. p-7118507
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B05385G 5 g
B053825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B05385G Supplier TCI America Supplier No. B05385G

Chemical Identifiers

CAS 2142-69-0
Molecular Formula C8H7BrO
Molecular Weight (g/mol) 199.05
MDL Number MFCD00000067
InChI Key PIMNFNXBTGPCIL-UHFFFAOYSA-N
Synonym 2'-bromoacetophenone, 1-2-bromophenyl ethanone, o-bromoacetophenone, ethanone, 1-2-bromophenyl, 1-acetyl-2-bromobenzene, 1-2-bromophenyl ethan-1-one, acetophenone, 2'-bromo, 1-2-bromomphenyl ethanone, acetophenone, o-bromo, o-bromophenyl methyl ketone
PubChem CID 75060
IUPAC Name 1-(2-bromophenyl)ethan-1-one
SMILES CC(=O)C1=CC=CC=C1Br

Specifications

Color Yellow
Boiling Point 136°C
Quantity 5 g
Formula Weight 199.05
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2′-Bromoacetophenone