missing translation for 'onlineSavingsMsg'
Learn More

2'-Bromoacetophenone, 99%

Catalog No. AAA1198209
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1198209 10 g
AAA1198218 50 g
AAA1198230 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1198209 Supplier Thermo Scientific Chemicals Supplier No. A1198209

CAS: 2142-69-0 | C8H7BrO | 199.05 g/mol

2-bromoacetophenone is employed in the selective derivatization of cytosine moieties for the determination of global DNA methylation by reversed phase high performance liquid chromatography with spectrofluorimetric detection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-bromoacetophenone is employed in the selective derivatization of cytosine moieties for the determination of global DNA methylation by reversed phase high performance liquid chromatography with spectrofluorimetric detection.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep away from oxidizing agents.

Chemical Identifiers

CAS 2142-69-0
Molecular Formula C8H7BrO
Molecular Weight (g/mol) 199.05
MDL Number MFCD00000067
InChI Key PIMNFNXBTGPCIL-UHFFFAOYSA-N
Synonym 2'-bromoacetophenone, 1-2-bromophenyl ethanone, o-bromoacetophenone, ethanone, 1-2-bromophenyl, 1-acetyl-2-bromobenzene, 1-2-bromophenyl ethan-1-one, acetophenone, 2'-bromo, 1-2-bromomphenyl ethanone, acetophenone, o-bromo, o-bromophenyl methyl ketone
PubChem CID 75060
IUPAC Name 1-(2-bromophenyl)ethanone
SMILES CC(=O)C1=CC=CC=C1Br

Specifications

Density 1.476
Boiling Point 116°C to 117°C (12 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.568
Quantity 10 g
Beilstein 1931534
Solubility Information Insoluble in water.
Formula Weight 199.05
Percent Purity 99%
Chemical Name or Material 2'-Bromoacetophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 218-398-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only