missing translation for 'onlineSavingsMsg'
Learn More

2'-Bromoacetophenone, 98+%

Catalog No. p-4509496
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC179760050 5 g Glass bottle
AC179760250 25 g Glass bottle
AC179761000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC179760050 Supplier Thermo Scientific Chemicals Supplier No. 179760050

CAS: 2142-69-0 | C8H7BrO | 199.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2142-69-0
Molecular Formula C8H7BrO
Molecular Weight (g/mol) 199.05
MDL Number MFCD00000067
InChI Key PIMNFNXBTGPCIL-UHFFFAOYSA-N
Synonym 2'-bromoacetophenone, 1-2-bromophenyl ethanone, o-bromoacetophenone, ethanone, 1-2-bromophenyl, 1-acetyl-2-bromobenzene, 1-2-bromophenyl ethan-1-one, acetophenone, 2'-bromo, 1-2-bromomphenyl ethanone, acetophenone, o-bromo, o-bromophenyl methyl ketone
PubChem CID 75060
IUPAC Name 1-(2-bromophenyl)ethanone
SMILES CC(=O)C1=CC=CC=C1Br

Specifications

Color Orange to Yellow
Density 1.4760g/mL
Boiling Point 116.0°C to 117.0°C (12.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.5650 to 1.5690
Linear Formula BrC6H4COCH3
Quantity 5 g
Specific Gravity 1.476
Solubility Information Solubility in water: practically insoluble in water. Other solubilities: freely soluble in alcohol,benzene,chlorofom,eth
Formula Weight 199.05
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 2'-Bromoacetophenone
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 218-398-1

RUO – Research Use Only