missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoacetophenone, 98%

Catalog No. AAA1557636
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1557636 500 g
AAA1557614 25 g
AAA1557622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1557636 Supplier Thermo Scientific Chemicals Supplier No. A1557636
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 70-11-1 | C8H7BrO | 199.047 g/mol

It is used in preparation of crystalline esters from acids. 2-Bromoacetophenone is a complete and irreversible inactivator of human liver aldehyde dehydrogenase isoenzymes E1 and E2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in preparation of crystalline esters from acids. 2-Bromoacetophenone is a complete and irreversible inactivator of human liver aldehyde dehydrogenase isoenzymes E1 and E2.

Solubility
Insoluble in water.

Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent, acids & bases.

Chemical Identifiers

CAS 70-11-1
Molecular Formula C8H7BrO
Molecular Weight (g/mol) 199.047
MDL Number MFCD00000195
InChI Key LIGACIXOYTUXAW-UHFFFAOYSA-N
Synonym 2-bromoacetophenone, phenacyl bromide, 2-bromo-1-phenylethan-1-one, bromoacetophenone, ethanone, 2-bromo-1-phenyl, bromomethyl phenyl ketone, alpha-bromoacetophenone, stauffer 4644, omega-bromoacetophenone, acetophenone, 2-bromo
PubChem CID 6259
ChEBI CHEBI:51846
IUPAC Name 2-bromo-1-phenylethanone
SMILES C1=CC=C(C=C1)C(=O)CBr

Specifications

Melting Point 48°C to 51°C
Density 1.65
Boiling Point 140°C (11 mmHg)
Flash Point >110°C (230°F)
Odor Pungent
Quantity 500 g
UN Number UN2645
Beilstein 606474
Merck Index 14,1402
Solubility Information Insoluble in water.
Formula Weight 199.05
Percent Purity 98%
Chemical Name or Material 2-Bromoacetophenone
Show More Show Less
Hazard Category H301+H331-H314
Hazard Statement GHS H Statement
H331-H314-H318
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: PHENACYL BROMIDE
EINECSNumber 200-724-9
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only