missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2'-Bromoacetanilide 98.0+%, TCI America™
Supplier: TCI America B313925G
Specifications
| 2′-Bromoacetanilide | |
| 102°C | |
| C8H8BrNO | |
| 25 g | |
| VOBKUOHHOWQHFZ-UHFFFAOYSA-N | |
| N-(2-bromophenyl)acetamide | |
| 136416 | |
| ≥98.0% (GC) |
| 614-76-6 | |
| White-Yellow | |
| MFCD00099252 | |
| n-2-bromophenyl acetamide, 2'-bromoacetanilide, o-bromoacetanilide, acetamide, n-2-bromophenyl, acetanilide, 2'-bromo, n-2-bromo-phenyl-acetamide, ortho-bromoacetanilide, 2/'-bromoacetanilide, acmc-209msx, 2-bromo-n-acetylaniline | |
| CC(=O)NC1=CC=CC=C1Br | |
| 214.06 | |
| 214.06 | |
| Crystalline Powder |
Chemical Identifiers
| 614-76-6 | |
| 214.06 | |
| VOBKUOHHOWQHFZ-UHFFFAOYSA-N | |
| 136416 | |
| CC(=O)NC1=CC=CC=C1Br |
| C8H8BrNO | |
| MFCD00099252 | |
| n-2-bromophenyl acetamide, 2'-bromoacetanilide, o-bromoacetanilide, acetamide, n-2-bromophenyl, acetanilide, 2'-bromo, n-2-bromo-phenyl-acetamide, ortho-bromoacetanilide, 2/'-bromoacetanilide, acmc-209msx, 2-bromo-n-acetylaniline | |
| N-(2-bromophenyl)acetamide |
Safety and Handling
TSCA : Yes