missing translation for 'onlineSavingsMsg'
Learn More

2'-Bromoacetanilide 98.0+%, TCI America™

Catalog No. B31395G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31395G 5 g
B313925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B31395G Supplier TCI America Supplier No. B31395G

Chemical Identifiers

CAS 614-76-6
Molecular Formula C8H8BrNO
Molecular Weight (g/mol) 214.06
MDL Number MFCD00099252
InChI Key VOBKUOHHOWQHFZ-UHFFFAOYSA-N
Synonym n-2-bromophenyl acetamide, 2'-bromoacetanilide, o-bromoacetanilide, acetamide, n-2-bromophenyl, acetanilide, 2'-bromo, n-2-bromo-phenyl-acetamide, ortho-bromoacetanilide, 2/'-bromoacetanilide, acmc-209msx, 2-bromo-n-acetylaniline
PubChem CID 136416
IUPAC Name N-(2-bromophenyl)acetamide
SMILES CC(=O)NC1=CC=CC=C1Br

Specifications

Melting Point 102°C
Color White-Yellow
Quantity 5 g
Formula Weight 214.06
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2′-Bromoacetanilide