missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoacetamide, 98%

Catalog No. p-71515
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC291100050 5 g Glass bottle
AC291100250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC291100050 Supplier Thermo Scientific Chemicals Supplier No. 291100050
Only null left

CAS: 683-57-8 | C2H4BrNO | 137.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 683-57-8
Molecular Formula C2H4BrNO
Molecular Weight (g/mol) 137.96
MDL Number MFCD00008025
InChI Key JUIKUQOUMZUFQT-UHFFFAOYSA-N
Synonym bromoacetamide, acetamide, 2-bromo, 2-bromo-acetamide, alpha-bromoacetamide, bromacetamide, bromoacetoamide, bromo acetamide, 2-bromoacetoamide, 2-bromo acetamide, sjyhcabiktp@
PubChem CID 69632
IUPAC Name 2-bromoacetamide
SMILES C(C(=O)N)Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 88°C to 91°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula BrCH2CONH2
Quantity 5 g
Beilstein 02, 216
Formula Weight 137.96
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromoacetamide, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Do not breat
RTECSNumber AB4587000
TSCA TSCA

RUO – Research Use Only